missing translation for 'onlineSavingsMsg'
Learn More

Fadolmidine Hydrochloride, TRC

Product Code. 30407345
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407345 100 mg
30398883 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407345 Supplier Toronto Research Chemicals Supplier No. TRCF101040100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189353-32-0
Molecular Formula C13H15ClN2O
Molecular Weight (g/mol) 250.72
Synonym 2,3-Dihydro-3-(1H-imidazol-5-ylmethyl)-1H-Inden-5-ol Hydrochloride (1:1), 4-(6-Hydroxyindan-1-ylmethyl)-1H-imidazole Monohydrochloride, 4-(6-Hydroxyindan-1-ylmethyl)-1H-imidazolium Chloride, MPV 2426
SMILES Cl.Oc1ccc2CCC(Cc3c[nH]cn3)c2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H14N2O.ClH/c16-12-4-3-9-1-2-10(13(9)6-12)5-11-7-14-8-15-11;/h3-4,6-8,10,16H,1-2,5H2,(H,14,15);1H
Formula Weight 250.0873
Chemical Name or Material Fadolmidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.