Learn More
FADS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14118-1-AP-150UL
 View more versions of this product
Product Code. 16842614
  • 410.00€ / 150µL
Description
FLAD1 encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme.
Specifications
Specifications
FADS
Polyclonal
Unconjugated
FLAD1
FAD pyrophosphorylase, FAD synthase, FAD1, FADS, FLAD1, FMN adenylyltransferase, PP591, RP11 307C12.7
Rabbit
Antigen Affinity Chromatography
RUO
319945, 751787, 80308
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8NFF5, Q8R123
Flad1
FADS Fusion Protein Ag5267
150 μL
Primary
Human, Mouse, Rat