missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14118-1-AP-20UL
This item is not returnable.
View return policy
Description
FLAD1 encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme.Specifications
| FADS | |
| Polyclonal | |
| Unconjugated | |
| FLAD1 | |
| FAD pyrophosphorylase, FAD synthase, FAD1, FADS, FLAD1, FMN adenylyltransferase, PP591, RP11 307C12.7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 319945, 751787, 80308 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8NFF5, Q8R123 | |
| Flad1 | |
| FADS Fusion Protein Ag5267 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction