missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF1 Antibody (CPTC-FAF1-2), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08227V
This item is not returnable.
View return policy
Description
FAF1 Monoclonal antibody specifically detects FAF1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications
FAF1 | |
Monoclonal | |
DyLight 405 | |
CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1 | |
Recombinant human full-length FAF1 protein (Uniprot: Q9UNN5) | |
100 μg | |
Apoptosis, Cancer | |
11124 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
CPTC-FAF1-2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction