Learn More
FAM119A Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503803S
Description
FAM119A Monoclonal antibody specifically detects FAM119A in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| FAM119A | |
| Monoclonal | |
| 0.91 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15848 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9370 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| OTI2D3 | |
| Unconjugated | |
| ADIPOQ | |
| 2310038H17Rik; AI464204; Fam119a; family with sequence similarity 119, member A; HCA557b; heat shock protein 70kDa lysine (K) methyltransferase; hepatocellular carcinoma-associated antigen 557b; HSPA lysine methyltransferase; HSPA-KMT; LOC102553119; methyltransferase like 21A; methyltransferase like 21A; protein N-lysine methyltransferase METTL21A; methyltransferase-like protein 21A; methyltransferase-like protein 21A; protein N-lysine methyltransferase METTL21A; Mettl21A; Protein N-lysine methyltransferase METTL21A; protein N-lysine methyltransferase METTL21A-like; protein-lysine methyltransferase METTL21A | |
| Human recombinant protein fragment corresponding to amino acids 19-244 of human ADIPOQ produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.