missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM125A Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€ - 483.00€
Specifications
| Antigen | FAM125A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388476
|
Novus Biologicals
NBP3-03413-20UL |
20 μL |
204.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18332884
|
Novus Biologicals
NBP3-03413-100UL |
100 μL |
483.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM125A Polyclonal specifically detects FAM125A in Mouse samples. It is validated for Western Blot.Specifications
| FAM125A | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 93343 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| CFBP, CIN85/CD2AP family binding protein, CIN85/CD2AP family-binding protein, ESCRT-I complex subunit MVB12A, family with sequence similarity 125, member A, FLJ32495, multivesicular body subunit 12A, Protein FAM125A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-273 of human FAM125A (NP_612410.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title