Learn More
RPL10A Antibody (OTI4B2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44768
Product Code. 30306207
  • 735.00€ / 100µg
Description
RPL10A Monoclonal antibody specifically detects RPL10A in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPL10A
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
Primary
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
OTI4B2
Western Blot 1:500, Immunohistochemistry 1:2000
CSA-19, Csa-19,60S ribosomal protein L10a, NEDD6, NEDD-6, Neural precursor cell expressed developmentally down-regulated protein 6, ribosomal protein L10a
Full length human recombinant protein of human RPL10A (NP_009035) produced in E.coli.
100 μg
Core ESC Like Genes, Stem Cell Markers
4736
Human, Mouse, Rat
Antibody
IgG2a