missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM219B Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26092PCP
This item is not returnable.
View return policy
Description
FAM219B Polyclonal antibody specifically detects FAM219B in Human samples. It is validated for Western Blot, Immunocytochemistry
Specifications
| FAM219B | |
| Polyclonal | |
| Western Blot, Immunocytochemistry | |
| C15orf17, chromosome 15 open reading frame 17, FLJ00005, hypothetical protein LOC57184 | |
| Produced in rabbits immunized with E. coli-derived Human FAM219B fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 57184 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction