Learn More
FAM40B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25163-1-AP-20UL
 View more versions of this product
Product Code. 16863715
  • 145.00€ / 20µL
Description
Product No. 17293-1-AP specifically recognizes the 96 kDa protein.
The function of this protein has not been specifically defined.
Specifications
Specifications
FAM40B
Polyclonal
Unconjugated
STRIP2
FAM40B, KIAA1170, Protein FAM40B, STRIP2
Rabbit
Antigen Affinity Chromatography
RUO
320609, 57464
-20°C
Liquid
Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8C9H6, Q9ULQ0
Strip2
FAM40B Fusion Protein Ag18230
20 μL
Primary
Human, Mouse