missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM83C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | FAM83C |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656002
|
Novus Biologicals
NBP2-98186-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679904
|
Novus Biologicals
NBP2-98186-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
FAM83C Polyclonal antibody specifically detects FAM83C in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifica
| FAM83C | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C20orf128, Chromosome 20 Open Reading Frame 128, dJ614O4.7, EC 1.6.5, EC 6.3.5.3, Family With Sequence Similarity 83, Member C, Protein FAM83C | |
| Produced in rabbits immunized with E. coli-derived Human FAM83C fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 128876 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto