missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM90A1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99202AF532
This item is not returnable.
View return policy
Description
FAM90A1 Polyclonal antibody specifically detects FAM90A1 in Human samples. It is validated for Western Blot
Specifications
| FAM90A1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55138 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| family with sequence similarity 90, member A1, FLJ10408, hypothetical protein LOC55138 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human FAM90A1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction