missing translation for 'onlineSavingsMsg'
Learn More

Famotidine Sulfoxide, Mikromol™

Product Code. 30052142
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052142 100 mg 100mg
30052338 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052142 Supplier Mikromol Supplier No. MM0029.04
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Famotidine Sulfoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Famotidine Sulfoxide
CAS 90237-03-9
Analyte or Component Names Famotidine Sulfoxide
Shipping Condition Ice pack (-20°C)
Molecular Formula C8 H15 N7 O3 S3
Certifications/Compliance ISO 9001
Synonym 3-[2-(Diaminomethyleneamino)-1,3-thiazol-4-ylmethylsulphinyl]-N-sulphamoylpropanamidineFamotidine Sulfoxide, Famotidine Sulfoxide
InChI Formula InChI=1S/C8H15N7O3S3/c9-6(15-21(12,17)18)1-2-20(16)4-5-3-19-8(13-5)14-7(10)11/h3H,1-2,4H2,(H2,9,15)(H2,12,17,18)(H4,10,11,13,14)
SMILES NC(=Nc1nc(CS(=O)CCC(=NS(=O)(=O)N)N)cs1)N
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfinyl]-N'-sulfamoylpropanimidamide
Molecular Weight (g/mol) 353.44
Quantity 100 mg
Formula Weight 353.0398 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.