missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2 Antibody (1290C), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-61930F
This item is not returnable.
View return policy
Description
FANCD2 Monoclonal antibody specifically detects FANCD2 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDown
Specifications
| FANCD2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, KnockDown | |
| 1290C | |
| Western Blot, Knockout Validated | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| This FANCD2 Antibody (1290C) was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| 2177 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction