missing translation for 'onlineSavingsMsg'
Learn More

FANCG Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NB100-2566SS

 View more versions of this product

Product Code. 18048633

  • 213.00€ / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FANCG Polyclonal specifically detects FANCG in Human samples. It is validated for Western Blot, Immunoblotting.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FANCG
Polyclonal
Western Blot 1:1000, Immunoblotting reported in scientific literature (PMID 24910428)
O15287
FANCG
A peptide corresponding to the C-terminus of human FANCG (within amino acids 550-622). [SwissProt# O15287]
Unpurified
RUO
Primary
Human. Immunogen sequence has 88% homology to mouse.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunoblot
Unconjugated
Whole antisera with No Preservative
DNA repair protein XRCC9, FAG, Fanconi anemia group G protein, Fanconi anemia, complementation group G, Protein FACG, X-ray repair, complementing defective, in Chinese hamster, 9, XRCC9X-ray repair complementing defective repair in Chinese hamster cells 9
Rabbit
69 kDa
0.025 mL
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Genes Sensitive to DNA Damaging Agents
2189
Human
Antisera
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only