missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAPP2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518211
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000. Preliminary analysis showed no signal in Western Blot.
Cargo transport protein that is required for apical transport from the Golgi complex. Transports AQP2 from the trans-Golgi network (TGN) to sites of AQP2 phosphorylation. Mediates the non-vesicular transport of glucosylceramide (GlcCer) from the trans-Golgi network (TGN) to the plasma membrane and plays a pivotal role in the synthesis of complex glycosphingolipids. Binding of both phosphatidylinositol 4-phosphate (PIP) and ARF1 are essential for the GlcCer transfer ability. Also required for primary cilium formation, possibly by being involved in the transport of raft lipids to the apical membrane, and for membrane tubulation.
Specifications
| FAPP2 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHA8 | |
| 6330400G01; AA517676; BC052360; FAPP2; Fapp-2; hFAPP2; MGC3358; PH domain-containing family A member 8; phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; pleckstrin homology domain containing A8; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8; pleckstrin homology domain-containing family A member 8; PLEKHA8; RGD1563940; Serologically defined breast cancer antigen NY-BR-86 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 84725 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96JA3 | |
| PLEKHA8 | |
| Synthetic peptide sequence (DIQTALRNPTENT) corresponding to the C-terminus amino acids of PLEKHA8 (aa 428-434). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction