missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550411
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total FAR2 protein.
This gene belongs to the short chain dehydrogenase/reductase superfamily. It encodes a reductase enzyme involved in the first step of wax biosynthesis wherein fatty acids are converted to fatty alcohols. The encoded peroxisomal protein utilizes saturated fatty acids of 16 or 18 carbons as preferred substrates. Alternatively spliced transcript variants have been observed for this gene. Related pseudogenes have been identified on chromosomes 2, 14 and 22.
Specifications
| FAR2 | |
| Polyclonal | |
| Unconjugated | |
| FAR2 | |
| epididymis secretory protein Li 81; FAR2; fatty acyl CoA reductase 2; fatty acyl-CoA reductase 2; HEL-S-81; male sterility domain-containing protein 1; MLSTD1; SDR10E2; short chain dehydrogenase/reductase family 10E, member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 330450, 55711 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q7TNT2, Q96K12 | |
| FAR2 | |
| Fusion protein corresponding to a region derived from internal residues of human fatty acyl CoA reductase 2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction