Learn More
Epredia™ Lab Vision™ Fascin Ab-1, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Fascin Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1112-RQ
Additional Details : Weight : 0.01200kg
Description
Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predominantly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However, Reed Sternberg cells in Hodgkin's lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.Host Species: Mouse
Clone: FCN01; same as 55K-2
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Fascin purified from HeLa cells
Molecular Weight: 55 kDa
Positive Control: Hodgkin's lymphoma or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Fascin Ab-1 | |
Monoclonal | |
Unconjugated | |
Fascin purified from HeLa cells | |
IVD | |
Primary |
Immunohistochemistry (Paraffin) | |
FCN01; same as 55K-2 | |
Mouse | |
12 mL | |
Neuroscience | |
Ready-to-Use |