missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fascin Antibody (FSCN1/416), Alexa Fluor™ 594 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11582AF594
This item is not returnable.
View return policy
Description
Fascin Monoclonal antibody specifically detects Fascin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Fascin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| FAN1,55 kDa actin-bundling protein, fascin homolog 1, actin-bundling protein (Strongylocentrotus purpuratus), FLJ38511, HSN, p55actin bundling protein, singed (Drosophila)-like (sea urchin fascin homolog like), singed-like (fascin homolog, sea urchin), Singed-like protein, SNLfascin | |
| Full length recombinant human Fascin protein (Uniprot: Q16658) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 6624 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FSCN1/416 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction