Learn More
Invitrogen™ Fasinumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559669
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Fasinumab has been used in trials studying the basic science and treatment of Sciatica, Back Pain, Low Back Pain, Osteoarthritis, Hip, and Abdominal Pain Upper, among others.
Specifications
| Fasinumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| REGN475; SAR164877 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |