missing translation for 'onlineSavingsMsg'
Learn More

Fast Corinth V Salt, TRC

Product Code. 30396795
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396795 1 g
30390762 500 mg
30389123 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396795 Supplier Toronto Research Chemicals Supplier No. TRCF1033301G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61966-14-1
Molecular Formula 2(C15H14N5O3 ) . Cl4Zn
Molecular Weight (g/mol) 831.8
Synonym Diazol Dark Violet K, Fast Corinth V Zinc Chloride Double Salt, Bis{2-methoxy-5-methyl-4-[(4-methyl-2-nitrophenyl)diazenyl]benzenediazonium}tetrachlorozincate, Mono((E)-2-methoxy-5-methyl-4-((4-methyl-2-nitrophenyl)diazenyl)benzenediazonium) Monozinc(IV) Tetrachloride
SMILES [Cl-][Zn+2]([Cl-])([Cl-])[Cl-].COc1cc(N=Nc2ccc(C)cc2[N+](=O)[O-])c(C)cc1[N+]#N.COc3cc(N=Nc4ccc(C)cc4[N+](=O)[O-])c(C)cc3[N+]#N

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C15H14N5O3.4ClH.Zn/c2*1-9-4-5-11(14(6-9)20(21)22)18-19-12-8-15(23-3)13(17-16)7-10(12)2;;;;;/h2*4-8H,1-3H3;4*1H;/q2*+1;;;;;+2/p-4
Formula Weight 828.0239
Chemical Name or Material Fast Corinth V Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.