missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Favipiravir
GREENER_CHOICE

Product Code. 18717116
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18717116

Brand: Tocris Bioscience™ 7225/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Viral RNA polymerase inhibitor

Favipiravir is a viral RNA polymerase inhibitor; exhibits no effects on mammalian RNA polymerase. Prodrug that is phosphoribosylated by cellular enzymes to its active form, Favipiravir-ribofuranosyl-5'-triphosphate (RTP). Reduces infection rate of SARS-CoV-2 in Vero E6 cells in vitro (IC50 = 61.88 μM). Inhibits influenza virus RNA-dependent RNA polymerase (RdRP) activity (IC50 = 341 nM). Induces lethal mutagenesis in MDCK cells inoculated with seasonal influenza A (H1N1) viruses. Protects mice against Ebola virus.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Fluoro-3,4-dihydro-3-oxo-2-pyrazinecarboxamide
CAS 259793-96-9
Quantity 50 mg
Target RNA/DNA Polymerase Inhibitors
Molecular Formula C5H4FN3O2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.