missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
132.87€ - 359.47€
Specifications
| Antigen | FBF1 |
|---|---|
| Concentration | 0.26 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
This antibody is induced by the fragment (492-792aa) of FBF1. It recognizes isoform 1, 2, 3, 5 and 6.
Keratin-binding protein required for epithelial cell polarization. Involved in apical junction complex (AJC) assembly via its interaction with PARD3. Required for ciliogenesis.Specifications
| FBF1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| A2A870, Q8TES7 | |
| 217335, 287836, 85302 | |
| FBF1 Fusion Protein Ag1954 | |
| Primary | |
| -20°C | |
| FBF1 |
| 0.26 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Alb, albatross, Fas (TNFRSF6) binding factor 1, FBF 1, FBF1, FBF1 Ens, HCG1989313, isoform CRA_b, KIAA1863 | |
| Fbf1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title