Learn More
FBLIM1 Monoclonal Antibody (OTI3G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807282
Description
This gene encodes a protein with an N-terminal filamin-binding domain, a central proline-rich domain, and, multiple C-terminal LIM domains. This protein localizes at cell junctions and may link cell adhesion structures to the actin cytoskeleton. This protein may be involved in the assembly and stabilization of actin-filaments and likely plays a role in modulating cell adhesion, cell morphology and cell motility. This protein also localizes to the nucleus and may affect cardiomyocyte differentiation after binding with the CSX/NKX2-5 transcription factor. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| FBLIM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WUP2 | |
| FBLIM1 | |
| Human recombit protein fragment corresponding to amino acids 1-242 of human FBLIM1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3G1 | |
| Unconjugated | |
| FBLIM1 | |
| CAL, FBLP-1, FBLP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54751 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.