Learn More
FBXO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14590-1-AP-20UL
 View more versions of this product
Product Code. 16803304
  • 145.00€ / 20µL
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is highly similar to the rat NFB42 (neural F Box 42 kDa) protein which is enriched in the nervous system and may play a role in maintaining neurons in a postmitotic state.
Specifications
Specifications
FBXO2
Polyclonal
Unconjugated
FBXO2
F box only protein 2, F box protein 2, FBG1, Fbs1, FBX2, FBXO2, NFB42
Rabbit
Antigen Affinity Chromatography
RUO
230904, 26232, 85273
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80UW2, Q9UK22
Fbxo2
FBXO2 Fusion Protein Ag6122
20 μL
Primary
Rat, Human, Mouse