missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO8 Antibody (OTI3C5), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71794V
This item is not returnable.
View return policy
Description
FBXO8 Monoclonal specifically detects FBXO8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FBXO8 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FBXO8 | |
| Recombinant protein fragment corresponding to amino acids 1-231 of human FBXO8 (NP_036312) produced in E.coli. | |
| 0.1 mL | |
| 26269.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3C5 | |
| Western Blot, Immunohistochemistry | |
| F-box only protein 8, F-box protein 8, F-box/SEC7 protein FBS, FBSFBX8F-box protein Fbx8 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction