Learn More
FBXO8 Monoclonal Antibody (OTI1A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807553S
Description
FBXO8 Monoclonal antibody specifically detects FBXO8 in Human samples. It is validated for Western BlotSpecifications
| FBXO8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NRD0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26269 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1A9 | |
| Unconjugated | |
| Fbxo8 | |
| DC10; F-box only protein 8; F-box protein 8; F-box protein Fbx8; F-box/SEC7 protein FBS; FBS; Fbx8; FBXO8; UNQ1877/PRO4320 | |
| Human recombinant protein fragment corresponding to amino acids 1-231 of human FBXO8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.