missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXW2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11499-1-AP-20UL
This item is not returnable.
View return policy
Description
Two isoforms were produced by alternative splicing with predicted MW of 52 and 43 kDa. 11499-1-AP can recognize both forms.
F-box proteins are an expanding family of eukaryotic proteins characterized by an approximately 40 amino acid motif, the F box. Some F-box proteins have been shown to be critical for the ubiquitin-mediated degradation of cellular regulatory proteins. In fact, F-box proteins are one of the four subunits of ubiquitin protein ligases, called SCFs. SCF ligases bring ubiquitin conjugating enzymes to substrates that are specifically recruited by the different F-box proteins. Mammalian F-box proteins are classified into three groups based on the presence of either WD-40 repeats, leucine-rich repeats, or the presence or absence of other protein-protein interacting domains. This gene encodes the second identified member of the F-box gene family and contains multiple WD-40 repeats.Specifications
| FBXW2 | |
| Polyclonal | |
| Unconjugated | |
| FBXW2 | |
| FBW2, FBXW2, Fwd2, Md6, Protein MD6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26190, 30050, 311881 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B2RZ17, Q60584, Q9UKT8 | |
| Fbxw2 | |
| FBXW2 Fusion Protein Ag2045 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction