missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RII/CD32 Antibody (FCGR2A/479), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08220AF532
This item is not returnable.
View return policy
Description
Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| Fc gamma RII/CD32 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| Recombinant human Fc gamma RII/CD32 protein (Uniprot: P12318) | |
| 100 μg | |
| Immunology | |
| 2213 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence | |
| FCGR2A/479 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction