missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCER1G Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115222
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FCER1G.
The high affinity IgE receptor is a key molecule involved in allergic reactions. It is a tetramer composed of 1 alpha, 1 beta, and 2 gamma chains. The gamma chains are also subunits of other Fc receptors.
Specifications
| FCER1G | |
| Polyclonal | |
| Unconjugated | |
| FCER1G | |
| AI573376; CD23; Fc epsilon receptor Ig; Fc fragment of IgE receptor Ig; Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide; Fc receptor gamma-chain; Fc receptor, IgE, high affinity I, gamma polypeptide; Fce1g; fc-epsilon RI-gamma; FcepsilonRI; FCER1G; FceRI gamma; FcR[g]; FCRG; FcRgamma; FcR-gamma; gamma polypeptide; high affinity immunoglobulin epsilon receptor subunit gamma; IgE Fc receptor subunit gamma; immunoglobulin E receptor, high affinity, gamma chain; Ly-50 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14127, 2207, 25441 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P20411, P20491, P30273 | |
| FCER1G | |
| A synthesized peptide derived from human FCER1G(Accession P30273), corresponding to amino acid residues K52-Q86. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction