missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLB/FCRY Antibody (454217), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4705L
This item is not returnable.
View return policy
Description
FCRLB/FCRY Monoclonal antibody specifically detects FCRLB/FCRY in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| FCRLB/FCRY | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 127943 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 454217 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FCRL2FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2FcRY, RP11-474I16.6 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRLB/FCRY, Ala18-Ser426, Accession # Q6BAA4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction