missing translation for 'onlineSavingsMsg'
Learn More

(-)-Fenchone, 98+%

Product Code. 11328176
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g

Product Code. 11328176

Brand: Thermo Scientific Alfa Aesar L15151.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

CAS: 7787-20-4 | C10H16O | 152.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-20-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00151104
InChI Key LHXDLQBQYFFVNW-GMSGAONNSA-N
Synonym l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
PubChem CID 3034206
IUPAC Name (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C

Specifications

Melting Point 3°C to 5°C
Density 0.945
Boiling Point 192°C to 194°C
Flash Point 73°C (163°F)
Refractive Index 1.463
Quantity 250 g
Beilstein 2042710
Optical Rotation −52° (Neat)
Formula Weight 152.24
Percent Purity ≥98%
Chemical Name or Material (-)-Fenchone
Show More Show Less
compliance-icons
Product Identifier
  • (-)-Fenchone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : (-)-Fenchone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.