missing translation for 'onlineSavingsMsg'
Learn More

Fenofibrate, Tocris Bioscience™

Product Code. 18014992
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18014992 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18014992 Supplier Tocris Bioscience Supplier No. 4113/50

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

PPARα agonist

PPARα agonist; hypolipidemic. Attenuates insulin resistance and increases high-density lipoprotein-cholesterol in db/db mice.
TRUSTED_SUSTAINABILITY

Specifications

CAS 49562-28-9
Molecular Formula C20H21ClO4
Quantity 50 mg
Synonym fenofibrate, procetofen, antara, lipanthyl, lipantil, tricor, fenobrate, lipidil, secalip, fenoglide
InChI Key YMTINGFKWWXKFG-UHFFFAOYSA-N
SMILES CC(C)OC(=O)C(C)(C)OC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl
IUPAC Name propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
Molecular Weight (g/mol) 360.834
PubChem CID 3339
ChEBI CHEBI:5001
Formula Weight 360.83
Percent Purity >99%
Chemical Name or Material Fenofibrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.