missing translation for 'onlineSavingsMsg'
Learn More

Fenthoxon, TRC

Product Code. 30040821
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040821 100 mg 100mg
30040442 10 mg 10mg
30011804 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040821 Supplier Toronto Research Chemicals Supplier No. TRCF279000100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6552-12-1
Molecular Formula C10 H15 O4 P S
Molecular Weight (g/mol) 262.26
Synonym Phosphoric acid, dimethyl 3-methyl-4-(methylthio)phenyl ester, Phosphoric acid, dimethyl 4-(methylthio)-m-tolyl ester (7CI,8CI), Bayoxon, Fenoxon, Fenthion oxon, Fenthoxon, Fenthion-oxon, Fenthion Oxon
IUPAC Name dimethyl (3-methyl-4-methylsulfanylphenyl) phosphate
SMILES COP(=O)(OC)Oc1ccc(SC)c(C)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H15O4PS/c1-8-7-9(5-6-10(8)16-4)14-15(11,12-2)13-3/h5-7H,1-4H3
Formula Weight 262.0429
Percent Purity >95%
Chemical Name or Material Fenthoxon
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.