Learn More
Fenticonazole nitrate, Thermo Scientific™
Product Code. 17595545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17595545 1 g 1g
17505555 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 17595545 Supplier Thermo Scientific Acros Supplier No. 468790010
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Chemical Identifiers
CAS 73151-29-8
Molecular Formula C24H21Cl2N3O4S
Molecular Weight (g/mol) 518.41
MDL Number 00941391
InChI Key FJNRUWDGCVDXLU-UHFFFAOYNA-N
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-{[4-(phenylsulfanyl)phenyl]methoxy}ethyl]-1H-imidazole; nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(C(CN2C=CN=C2)OCC2=CC=C(SC3=CC=CC=C3)C=C2)C(Cl)=C1
Specifications
Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Crystalline Powder
Color White
Melting Point 133°C to 137°C
Merck Index 13-4033
Quantity 1 g
Chemical Name or Material Fenticonazole nitrate