Learn More
Chemetrics™ Ferric Thiocyanate Method Iron in Brine CHEMets Refill
Product Code. 12073313
Change view
Click to view available options
Range:
0 to 100 and 1 to 100 to 1000 mg/L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
12073313 0 to 100 and 1 to 100 to 1000 mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 12073313 Supplier Chemetrics™ Supplier No. R6002
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Specifications
For Use With (Application) High concentrations in surface waters can indicate the presence of industrial effluents or runoff.
For Use With (Equipment) CHEMets Kit
No. of Tests 30
Quantity 1 Each
Range 0 to 100 and 1 to 100 to 1000 mg/L
Test Method Ferric Thiocyanate
Type Iron in Brine CHEMets Refill