Learn More
Chemetrics™ Ferric Thiocyanate Method Iron in Brine CHEMets Refill
Specifications
Specifications
| For Use With (Application) | High concentrations in surface waters can indicate the presence of industrial effluents or runoff. |
| For Use With (Equipment) | CHEMets Kit |
| No. of Tests | 30 |
| Quantity | 1 Each |
| Range | 0 to 100 and 1 to 100 to 1000 mg/L |
| Test Method | Ferric Thiocyanate |
| Type | Iron in Brine CHEMets Refill |