missing translation for 'onlineSavingsMsg'
Learn More

Ferulic Acid Methyl Ester, TRC

Product Code. 30133326
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133326 25 g 25g
30132460 5 g 5g
30007735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133326 Supplier Toronto Research Chemicals Supplier No. TRCF30894025G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2309-07-1
Molecular Formula C11 H12 O4
Molecular Weight (g/mol) 208.21
Synonym 2-Propenoic acid, 3-(4-hydroxy-3-methoxyphenyl)-, methyl ester, Cinnamic acid, 4-hydroxy-3-methoxy-, methyl ester (6CI,7CI,8CI), 3-(4-Hydroxy-3-methoxyphenyl)-2-propenoic acid methyl ester, Ferulic acid methyl ester, Methyl 4'-hydroxy-3'-methoxycinnamate, Methyl ferulate, NSC 74548
IUPAC Name methyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
SMILES COC(=O)\C=C\c1ccc(O)c(OC)c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C11H12O4/c1-14-10-7-8(3-5-9(10)12)4-6-11(13)15-2/h3-7,12H,1-2H3/b6-4+
Formula Weight 208.0736
Chemical Name or Material Ferulic acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.