Learn More
Cytiva Fetal Bovine Serum (U.S.), Super Low IgG
Product Code. 11547310
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated, 25 to 40 doses
None
Unit Size:
100mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
11547310 100 mL None 100mL
16221122 500 mL Irradiated, 25 to 40 doses 500mL
12360143 500 mL Heat Inactivated 500mL
10329953 500 mL None 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11547310 Supplier Cytiva Supplier No. SH30898.02
Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production
Fetal bovine serum (FBS) has natural levels of IgG that are too high for some cell culture applications. Lots can be prescreened to find the lowest naturally occuring levels, but as a biological product, FBS is frequently in short supply. Even lots with naturally low IgG levels may still contain too much for some applications. Examples of typical IgG levels in bovine serum products can be found in the table below.
  • Performance is equivalent to non-processed FBS
  • Suitable for applications where low levels of specific antibody are required (e.g. propagation of bovine viruses, monoclonal antibody production)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2 to 8°C
Quantity 100 mL
Processing None
Heat Inactivated No