missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
241.00€ - 326.00€
Specifications
| Antigen | Fetuin |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634866
|
Novus Biologicals
NBP2-99828-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630217
|
Novus Biologicals
NBP2-99828-50ul |
50 μg |
241.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fetuin Polyclonal antibody specifically detects Fetuin in Human samples. It is validated for ELISASpecifications
| Fetuin | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| fetuin B, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
| Produced in rabbits immunized with purified, recombinant Human Fetuin (Accession#: NP_055190.2; Met 1-Pro 382) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 26998 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title