missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FFAR2/GPR43 Antibody (1000624) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10082L
This item is not returnable.
View return policy
Description
FFAR2/GPR43 Monoclonal antibody specifically detects FFAR2/GPR43 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| FFAR2/GPR43 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 1000624 | |
| Flow Cytometry, CyTOF-ready | |
| free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43FFA2R, G-protein coupled receptor 43 | |
| NS0 Mouse myeloma cell line transfected with human FFAR2/GPR43, Met1-Glu330, Accession # O15552 | |
| 0.1 mL | |
| GPCR | |
| 2867 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction