Learn More
Coverguard™ FFP2 NR D Face Mask
The Coverguard™ FFP2 masks are single-use and protect against non-toxic solid and liquid aerosols in applications such as farming, silviculture and fishing, light industries, maintenance, infrastructure and building, DIY, as well as general use in the lab or field.
207.00€ - 658.00€
Specifications
Certifications/Compliance (Europe) | CE PPE Category III FFP2 NR D, EN 149:2001 + A1:2009 |
---|---|
Disposable | Single-use |
For Use With (Equipment) | Chemical and Pharmaceutical Industries, Light Industries, Heavy and Process Industries, Construction Finishings, Building Industry, Infrastructure |
Resistant To | Non-Toxic Solid and Liquid Aerosols |
Sterility | Non-sterile |
Product Code | Brand | Style | Nosepiece Type | With Exhalation Valve | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Style | Nosepiece Type | With Exhalation Valve | Price | Quantity & Availability | |||
12073186
|
Coverguard™
23201 |
Shell Mask | Adjustable Nose Clip with PU Foam Nose Pad | No |
483.00€
Pack of 240 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||
12083186
|
Coverguard™
23205 |
Half Mask with Exhalation Valve | Adjustable Nose Clip | Yes |
658.00€
Pack of 240 |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||
12093186
|
Coverguard™
23206 |
Shell Mask with Exhalation Valve | Adjustable Nose Clip with PU Nose Pad | Yes |
207.00€
Pack of 120 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||
Description
- PPE Cat. III FFP2 NR D
- Disposable
- Electrostatic filter media
- Sliding elastic band for better adjustment
- Latex-free, non-woven polypropylene
- Adjustable nose clip
Specifications
CE PPE Category III FFP2 NR D, EN 149:2001 + A1:2009 | |
Chemical and Pharmaceutical Industries, Light Industries, Heavy and Process Industries, Construction Finishings, Building Industry, Infrastructure | |
Non-sterile |
Single-use | |
Non-Toxic Solid and Liquid Aerosols |