Learn More
Coverguard™ FFP2 NR D SL Half Mask with Exhalation Valve
Product Code. 12063196
Change view
Click to view available options
Resistant To:
Aerosols, Low Toxic Dust
Unit Size:
Pack of 120
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Resistant To unitSize
12063196 Aerosols, Low Toxic Dust Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12063196 Supplier Coverguard™ Supplier No. 23276
The preformed design of the Disposable FFP2 NR D SL Half Mask fits with glasses or goggles to avoid the risk of condensation. The ergonomic form offers an optimal adjustment to the face, without a nasal clip. Breathing resistance is reduced with the exhalation valve.
  • FFP2 NR D PPE Category III
  • Preformed design fits with glasses or goggles
  • Ergonomic form offers optimal fit
  • Exhalation valve reduces breathing resistance
Specifications
Certifications/Compliance (Europe) CE PPE Cat. III, FFP2 NR D, EN 149:2001 + A1:2009
Description FFP2 Preformed Shell Mask with Valve
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP2
With Exhalation Valve Yes
Resistant To Aerosols, Low Toxic Dust