missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98997-50ul
This item is not returnable.
View return policy
Description
FGF-10 Polyclonal antibody specifically detects FGF-10 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| FGF-10 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 4-8 uL/mg of lysate, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, produced by fibroblasts of urinary bladder lamina propria | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human FGF-10. | |
| 50 μg | |
| Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
| 2255 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction