missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-21 Antibody (CL6491) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76496AF594
This item is not returnable.
View return policy
Description
FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for Western Blot
Specifications
| FGF-21 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| FGF-21, fibroblast growth factor 21 | |
| This antibody was developed using a synthetic peptide derived from Q9NSA1, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 26291 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL6491 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction