Learn More
FGF21 Monoclonal Antibody (OTI2F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502868S
Description
FGF21 Monoclonal antibody specifically detects FGF21 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| FGF21 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75688 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5495 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI2F8 | |
| Unconjugated | |
| PPM1B | |
| FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196 | |
| Full length human recombinant protein of human PPM1B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.