missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114734
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FGF8.
Plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. Required for normal brain, eye, ear and limb development during embryogenesis. Required for normal development of the gonadotropin-releasing hormone (GnRH) neuronal system. Plays a role in neurite outgrowth in hippocampal cells.
Specifications
| FGF8 | |
| Polyclonal | |
| Unconjugated | |
| FGF8 | |
| AIGF; androgen-induced; Androgen-induced growth factor; FGF; FGF8; Fgf-8; Fibroblast growth factor; Fibroblast growth factor 8; fibroblast growth factor 8 (androgen-induced); HBGF-8; Heparin-binding growth factor 8; HH6; KAL6; MGC149376 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14179, 2253, 29349 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P37237, P55075 | |
| FGF8 | |
| A synthesized peptide derived from human FGF8(Accession P55075), corresponding to amino acid residues L152-T202. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction