Learn More
FGFR2 Antibody (98739) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB6843JF585
Product Code. 30164000
  • 639.00€ / 0.10mL
Description
FGFR2 Monoclonal antibody specifically detects FGFR2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Neutralization,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGFR2
Monoclonal
Janelia Fluor 585
BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms
RUO
Primary
Human
Liquid
Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF
98739
50mM Sodium Borate
Mouse
0.1 mL
Angiogenesis, Protein Kinase
2263
Store at 4°C in the dark.