Learn More
FGL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11827-1-AP-150UL
 View more versions of this product
Product Code. 16809163
  • 410.00€ / 150µL
Description
Fibrinogen-like protein 2 (Fgl2), also known as fibroleukin, is secreted by T cells and is involved in diseases in which thrombosis plays a pivotal role,such as virus-induced fulmit hepatitis, fetal loss syndrome and xenograft rejection. Constitutively xpressed in cytotoxic T cells, Fgl2 exerts immunosuppressive effects on both T cell proliferation and dendritic cell maturation.Fgl2 is a serine protease and directly cleaves prothrombin to thrombin. Fgl2 functions in the pathogenesis of diseases including viral-induced hepatitis and Th1 cytokine-induced fetal loss syndrome.
Specifications
Specifications
FGL2
Polyclonal
Unconjugated
FGL2
FGL2, FGL-2, fibrinogen like 2, Fibrinogen like protein 2, Fibroleukin, pT49, T49
Rabbit
Antigen Affinity Chromatography
RUO
10875, 14190, 84586
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P12804, Q14314
Fgl2
FGL2 Fusion Protein Ag2395
150 μL
Primary
Human, Mouse, Rat