Learn More
Thermo Scientific™ Fiberlite™ F21-48 x 2 Fixed-Angle Rotor
GREENER_CHOICE
Product Code. 10707845
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10707845 Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10707845 Supplier Thermo Scientific™ Supplier No. 75003664
  • ONLINE EXCLUSIVE PRICE
    3710.00€
    3087.00€ / Pack of 1
    Save 623.00€
    16% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Support micro-volume protocols using the fixed angle rotor, provided with a biocontainment lid.
Support micro-volume protocols using the Thermo Scientific™ Fiberlite™ F21-48 x 2 Fixed-Angle Rotor.
Features:
Fiberlite Rotor:
  • Reduces run-times with higher G-forces achieved with lightweight carbon fiber rotors
  • Ensures structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Ordering Information:
Tube adapters are sold separately in sets of 24; O-ring Refresher Kit (021-489-021) is also available. Contact your sales representative for information.
Compatible with:
Thermo Scientific Sorvall™ Legend™ X1, X3, Sorvall ST 40, Multifuge™ X1, X3, Megafuge 40, SL40 and Eppendorf™ 5810/5804 Series, Sorvall Legend T+, Multifuge 3+ Series, Sorvall Legend 23R, MR23i, Thermo Scientific™ Nalgene™ bottles and tubes
Note:
†Performance may vary by centrifuge model.
TRUSTED_SUSTAINABILITY
Specifications
Description Fiberlite F21-48 x 2 Fixed Angle Rotor for Thermo Scientific Sorvall Legend X1, X3, Sorvall ST 40
Type Fixed Angle Rotor
Capacity 48 x 2 mL
Max. RCF 25,055 x g
Max. Speed 15,200 rpm
Radius (Metric) Min. 5.1 cm
K Factor 455
Max. Tube Length 50 mm
Max. Tube Diameter 11 mm
For Use With (Equipment) Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge
Angle 45°
Includes Biocontainment lid
Biocontainment Yes