missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrin Antibody (UC45), Novus Biologicals™
Mouse Monoclonal Antibody
214.00€ - 372.00€
Specifications
| Antigen | Fibrin |
|---|---|
| Clone | UC45 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready |
| Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605198
|
Novus Biologicals
NBP2-50407-0.025ml |
0.025 mL |
214.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653818
|
Novus Biologicals
NBP2-50407 |
0.1 mL |
372.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibrin Monoclonal antibody specifically detects Fibrin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays, CyTOFSpecifications
| Fibrin | |
| 1 mg/mL | |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), CyTOF | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology | |
| PBS | |
| IgM | |
| Thiophilic Chromatography |
| UC45 | |
| Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Human acute monoblastic leukemia cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title