Learn More
Fibroblast activation protein alpha Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27596-1-AP-150UL
 View more versions of this product
Product Code. 18039895
  • 410.00€ / 150µL
Description
The Fibroblast activation protein alpha antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Fibroblast activation protein alpha. This antibody recognizes human, mouse antigen. The Fibroblast activation protein alpha antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
Fibroblast activation protein alpha
500 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunohistochemistry, ELISA
DPPIV, FAP, FAPA, Seprase
150 μL
2191
Store at -20°C. Stable for one year after shipment.